Orders LinkContactUsLinkComponent
Ultrafiltration vials
Ultrafiltration vials
Supplier:   Biotium
Ultrafiltration vials
Supplier:  Biotium
Certificates
Ultrafiltration vials
2 Options Available Below
Product Details & Documents
Ultrafiltration can be used to rapidly and easily remove small molecules such as Tris, glycerol and glycine from antibody solutions, or to concentrate antibodies prior to conjugation. They also can be used to remove free dye or free biotin from antibodies after conjugation.
For ultrafiltration of proteins, it is recommended to use a MWCO 3 to 6× smaller than the protein to be retained. Ultrafiltration vials with an MWCO of 3 kD are recommended for ultrafiltration of proteins larger than 10 kD. Vials with an MWCO of 10 kD are more suitable for ultrafiltration of antibodies and other proteins larger than 30 kD. Ultrafiltration is not effective for fractionating proteins (separating one protein from another protein based on size) unless there is at least a 10-fold difference in the protein molecular weights.
Product Family Options
Product Information
  • MWCO
    Availability
    Price
  • 22018.
    Ultrafiltration vials
    Ultrafiltration vials
    3 kD
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 96.00
    SFr. 96.00
    1 * 5 ST
       SFr. 96.00
  • 22004.
    Ultrafiltration vials
    Ultrafiltration vials
    10 kD
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 102.00
    SFr. 102.00
    1 * 5 ST
       SFr. 102.00