Order Entry
Switzerland
ContactUsLinkComponent
3-Nitro-2-picoline 97%
3-Nitro-2-picoline 97%
Catalog # ACRO453690050
CAS Number:  
undefined
3-Nitro-2-picoline 97%
Catalog # ACRO453690050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Light yellow to brown Liquid or low melting solid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %

Specification Test Results

About this item

Synonyms: 2-Methyl-3-nitropyridine
 :
 :138.1240 g/mol
Boiling Pt:104 °C (15 mmHg)
Flash Pt:83 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:18699-87-1
Formula:C₆H₆N₂O₂
MDL Number:MFCD04114137