Methyl arachidate 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Methyl eicosanoate, Methyl Arachidate
- CAS Number:1120-28-1
- Formula:C₂₁H₄₂O₂
- MDL Number:MFCD00009014
- Molecular Weight:326.56 g/mol
- Boiling Pt:215…216 °C (10 mmHg)
- Melting Pt:45…48 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: Methyl eicosanoate, Methyl Arachidate
- CAS Number:1120-28-1
- Formula:C₂₁H₄₂O₂
- MDL Number:MFCD00009014
- Molecular Weight:326.56 g/mol
- Boiling Pt:215…216 °C (10 mmHg)
- Melting Pt:45…48 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:45 to 48 °C
- GC:≥98.5 %
- Free acid:≤1 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



