Order Entry
Switzerland
ContactUsLinkComponent
 
L-Alanyl-L-glutamine
 
undefined
L-Alanyl-L-glutamine

 

  • CAS No.:
    39537-23-0
  • Storage conditions:
    Ambient
  • Form:
    White to off-white crystalline powder
  • MDL No.:
    MFCD00133046
  • Protein synonyms:
    H-Ala-Gln-OH
  • Protein/peptide name:
    L-Alanyl-L-glutamine
  • Purity:
    ≥96,0%
  • Molecular weight:
    217,23 g/mol
  • Molecular formula:
    C₈H₁₅N₃O₄

 

 

Frequently Bought Together