ContactUsLinkComponent
Sodium hydroxide, micropearls, extra pure
Corrosive
Sodium hydroxide, micropearls, extra pure
Certificates
undefined
Corrosive
Sodium hydroxide, micropearls, extra pure
Specification Test Results
  • Appearance:
    White Pearls
  • Particle size:
    20 to 40 mesh
  • Titration with HCl:
    ≥97.5 % (NaOH)