
Ammonium bromide 98%, extra pure
:12124-97-9
:
:H₄BrN
:MFCD00011419
:97.94 g/mol
:235 °C (760 mmHg)
:452 °C (subl.)
:2,42 g/cm³ (20 °C)
:GHS07
:Ambient
- Appearance:White Crystalline powder and/or chunks
- Titration Argentometric:≥97.5 %
- Sulfated ash:≤0.1 %
- Insoluble matter:≤0.01 % (in water)
Frequently Bought Together