Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Hydrochloric acid 37% for analysis (max. 0.01 ppm Hg)
  450550025.
 :  
undefined
 
 
Hydrochloric acid 37% for analysis (max. 0.01 ppm Hg)
  450550025.
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Residue after ignition:
    ≤0.0005 %
  • Titration with NaOH:
    ≥37.0 %
  • Chloride (Cl):
    ≤0.5 ppm
  • Phosphate (PO4):
    ≤0.5 ppm
  • Sulfite (SO3):
    ≤1 ppm
  • Sulfate (SO4):
    ≤1 ppm
  • Bromine (Br):
    ≤50 ppm
  • Aluminium (Al):
    ≤0.05 ppm
  • Ammonium (NH4):
    ≤1 ppm
  • Arsenic (As):
    ≤0.01 ppm
  • Barium (Ba):
    ≤0.02 ppm
  • Beryllium (Be):
    ≤0.02 ppm
  • Bismuth (Bi):
    ≤0.1 ppm
  • Cadmium (Cd):
    ≤0.01 ppm
  • Calcium (Ca):
    ≤0.5 ppm
  • Chromium (Cr):
    ≤0.02 ppm
  • Cobalt (Co):
    ≤0.01 ppm
  • Copper (Cu):
    ≤0.02 ppm
  • Germanium (Ge):
    ≤0.05 ppm
  • Iron (Fe):
    ≤0.2 ppm
  • Lead (Pb):
    ≤0.02 ppm
  • Lithium (Li):
    ≤0.01 ppm
  • Magnesium (Mg):
    ≤0.01 ppm
  • Manganese (Mn):
    ≤0.1 ppm
  • Mercury (Hg):
    ≤0.01 ppm
  • Molybdenum (Mo):
    ≤0.02 ppm
  • Nickel (Ni):
    ≤0.02 ppm
  • Potassium (K):
    ≤0.1 ppm
  • Sodium (Na):
    ≤0.5 ppm
  • Strontium (Sr):
    ≤0.01 ppm
  • Thallium (Tl):
    ≤0.05 ppm
  • Titanium (Ti):
    ≤0.1 ppm
  • Vanadium (V):
    ≤0.01 ppm
  • Zinc (Zn):
    ≤0.05 ppm
  • Zirconium (Zr):
    ≤0.1 ppm

 

 

 :
 :57 °C (760 mmHg)
 :–35 °C
 :1.18 g/cm³
 :Ambient
 :8,II
 :36.46 g/mol
 :1789
 :GHS05
 :GHS07
 :7647-01-0
 :HCl
 :MFCD00011324