Vanadium(V) oxide 99+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Number:1314-62-1
- Formula:O₅V₂
- MDL Number:MFCD00011457
- Molecular Weight:181.88 g/mol
- Boiling Pt:1750 °C (760 mmHg)
- Melting Pt:690 °C
- Density:3.35 g/cm³
- Storage Temperature:Ambient
- UN:2862
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Number:1314-62-1
- Formula:O₅V₂
- MDL Number:MFCD00011457
- Molecular Weight:181.88 g/mol
- Boiling Pt:1750 °C (760 mmHg)
- Melting Pt:690 °C
- Density:3.35 g/cm³
- Storage Temperature:Ambient
- UN:2862
- ADR:6.1,III
Specifications for Product Family
Specification Test Results
- Appearance:Dark yellow Powder
- Assay:≥99.0 %
- Particle size:≤1.2 mm
- Chromium (Cr):≤0.1 %
- Iron (Fe):≤0.1 %
- Phosphorus (P):≤0.04 %
- Potassium (K):≤0.4 %
- Silicon (Si):≤0.06 %
- Sodium (Na):≤0.3 %
- Sulfur (S):≤0.07 %



