Order Entry
Switzerland
ContactUsLinkComponent
 
 
Di-tert-butyl phosphonate 96%
 
undefined
 
Di-tert-butyl phosphonate 96%

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4190 to 1.4210 (20°C, 589 nm)
  • GC:
    ≥95.0 %

 

 

Frequently Bought Together