


Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Catalog # 448361000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2320
- Pk Info:4× 25 ml
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.005 % (Coulometric)
- GC:≥99.80 %
- Stabilizer:≥200 ppm BHT
- Residue after evaporation:≤0.001 %
- Filt.over 0.2 micron PFTE:Filtered
- Acidity (CH3COOH):≤0.0005 meq/g
- Peroxides:≤0.0025 % (H2O2)
Specification Test Results
About this item
:
:72.11 g/mol
Boiling Pt:66 °C (760 mmHg)
Melting Pt:–108.4 °C
Density:0.88 g/cm³
Flash Pt:–21 °C
UN:2056
ADR:3,II
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356