


Ethylene glycol monomethyl ether (2-methoxyethanol) ACS
:109-86-4
:
:C₃H₈O₂
:MFCD00002867
:76.1 g/mol
:124.6 °C (1013 hPa)
:–85 °C
:0.96 g/cm³
:38 °C
:1188
:3,III
:GHS07
:GHS08
:GHS02
:Ambient
- Appearance:Clear liquid
- Water:≤0.1 %
- GC:≥99.3 %
- Color scale:≤10 APHA
- Titrable acid:≤0.002 meq/g
Frequently Bought Together