Order Entry
Switzerland
ContactUsLinkComponent
 
Sodium hydrogen carbonate 99+% for HPLC
 
undefined
Sodium hydrogen carbonate 99+% for HPLC
 

 

  • Appearance:
    White Crystalline powder
  • UV:
    (1 M in water)
  • pH:
    6.5 to 8.5
  • Titration with HCl:
    ≥99.0 %

 

 

Frequently Bought Together