


Sodium methoxide 0.5 M in methanol, AcroSeal® ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:124-41-4
:
Formula:CH₃NaO
MDL Number:MFCD00012179
:54.02 g/mol
Boiling Pt:65 °C (760 mmHg)
Melting Pt:–98 °C
Density:0.8 g/cm³
Flash Pt:11 °C
UN:1992
ADR:3,II
:GHS06
:GHS08
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Solution
- Titration with HCl:0.48 to 0.52 M
- Clarity of solution:Passes test
Specification Test Results
Related Information
Frequently Bought Together