Order Entry
Switzerland
ContactUsLinkComponent
Hexane (mixture of isomers), technical
Flammable
Irritant
Health
Environment
Hexane (mixture of isomers), technical
Catalog # ACRO444340050
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
Hexane (mixture of isomers), technical
Catalog # ACRO444340050
CAS Number:  

Specifications

  • Pack type:
    Metal drum
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Refractive index:
    1.3770 to 1.3810 (20°C, 589 nm)
  • Density:
    0.664 to 0.678 kg/l

Specification Test Results

About this item

 :
 :86.18 g/mol
Boiling Pt:60 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.659 g/cm³
Flash Pt:–22 °C
Storage Temperature:Ambient
UN:1208
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520