Ethanol 99.8%, Technical Grade (not denatured)
Plastic drum, 5 L
Supplier: Thermo Fisher Scientific
Ethanol 99.8%, Technical Grade (not denatured)
Catalog # ACRO444220050
Supplier: Thermo Fisher Scientific
CAS Number: 64-17-5
Jump to:
Product Details & Documents
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Boiling Pt:78.3 °C (1013 hPa)
- Melting Pt:–114 °C
- Density:0.79 g/cm³
- Flash Pt:12 °C
- Storage Temperature:Ambient
- UN:1170
- ADR:3,II
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Refractive index:1.3600 to 1.3630 (20°C, 589 nm)
- Density:(20°C) 0.789 to 0.791 kg/l
- Ethanol:99.8 to 100.0 % v/v (calculated)
- GC:99.75 %
Specifications
- Pack type:Plastic drum
- Pk:5 L
- Cat. No.:ACRO444220050
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Safer Human and Environmental Health



