

Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-96-6
:
Formula:C₆H₁₄O₃
MDL Number:MFCD00008503
Boiling Pt:162 °C (760 mmHg)
Melting Pt:–64 °C
Density:0.937 g/cm³
Flash Pt:51 °C
UN:3271
ADR:3,III
:134.18 g/mol
:GHS08
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
Specification Test Results
Related Information
Frequently Bought Together