
Tetrakis(triphenylphosphine)palladium(0) 99.9% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14221-01-3
:
Formula:C₇₂H₆₀P₄Pd
MDL Number:MFCD00010012
:1155.58 g/mol
Melting Pt:103…107 °C (dec.)
Density:1 g/cm³ (20 °C)
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:Yellow to yellow-green Powder or crystals
- Infrared spectrum:Conforms
- Total trace metal impurities:≤0.1 %
- Palladium (Pd):8.8 to 9.6 %
Specification Test Results
Frequently Bought Together