
Boc-L-valine hydroxysuccinimide ester 98%
Catalog # ACRO440920050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Water:≤1.0 %
- HPLC:≥97.5 %
- Specific optical rotation:-36.0° to -40.0° (20°C, 589 nm) (c=1 in dioxane)
Specification Test Results
About this item
:
:314.338 g/mol
Melting Pt:126…129 °C
:GHS07
Storage Temperature:Freezer
CAS Number:3392-12-9
Formula:C₁₄H₂₂N₂O₆
MDL Number:MFCD00037906