N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207683-26-9
:
Formula:C₁₃H₂₀F₆N₃O₃P
MDL Number:MFCD00192457
:411.29 g/mol
Melting Pt:141…143.5 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Elemental Analysis:36.45 to 39.48 % (C)
Specification Test Results
Frequently Bought Together