
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:147804-30-6
:
Formula:C₁₁H₁₉NO₃
MDL Number:MFCD07779385
:213.28 g/mol
Melting Pt:58…59 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:55 to 60 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together