Molecular sieve A4 (0.4 nm, 4 Å), pellets Ø 3.2mm
:70955-01-0
:1318-02-1
:MFCD00131613
:1,1 to 2.2 g/cm³ (20 °C)
:Ambient
- Appearance:Beige Cylinders
- Bulk density:600 to 750 g/l
- Residual water:≤2 % (550°C, 2 h)
- Water adsorption:≥20 % (50% rH, 20°C)
Frequently Bought Together