
5-Iodopyridin-2-ol 97%
Catalog # ACRO436370010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:186 to 191 °C
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:221 g/mol
Melting Pt:186.7…190.1 °C
:GHS07
Storage Temperature:Ambient
CAS Number:13472-79-2
Formula:C₅H₄INO
MDL Number:MFCD00234058