



Antimony(V) chloride 1M in methylene chloride, AcroSeal®
Katalognr. 434521000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to dark yellow Solution
- Titration Argentometric:0.95 to 1.10 M
- Refractive index:1.5310 to 1.5350 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:299.02 g/mol
Boiling Pt:140 °C (1013 hPa)
Melting Pt:3 °C
Density:1.442 g/cm³
Storage Temperature:Ambient
UN:2922
ADR:8,II
:GHS05
:GHS07
:GHS08
:GHS09
CAS Number:7647-18-9
Formula:Cl₅Sb
MDL Number:MFCD00011213