ContactUsLinkComponent
1H-Tetrazole 3 - 4% (w/w) in acetonitrile, AcroSeal®
Flammable
Irritant
1H-Tetrazole 3 - 4% (w/w) in acetonitrile, AcroSeal®
Certificates
undefined
Flammable
Irritant
1H-Tetrazole 3 - 4% (w/w) in acetonitrile, AcroSeal®
Specification Test Results
  • Appearance:
    Clear colorless to light yellow Solution (May develop some turbidity or precipitate)
  • Water:
    ≤0.3 %
  • Strength:
    3.0 to 4.0 % (wt)
  • Titration with HClO4:
    31.0 to 35.0 %