Order Entry
Switzerland
ContactUsLinkComponent
HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98%, AcroSeal®
Toxic
Corrosive
Flammable
HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98%, AcroSeal®
Certificates
undefined
Toxic
Corrosive
Flammable
HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98%, AcroSeal®

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4060 to 1.4090 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Color scale:
    ≤10 APHA

Specification Test Results

Frequently Bought Together