Order Entry
Switzerland
ContactUsLinkComponent
 
Irritant
5-Fluoro-2-methylphenylboronic acid 97%
Catalog # ACRO430650050
CAS Number:  
undefined
Irritant
5-Fluoro-2-methylphenylboronic acid 97%
Catalog # ACRO430650050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    149 to 154 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
Melting Pt:116…118 °C
 :153.95 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:163517-62-2
Formula:C₇H₈BFO₂
MDL Number:MFCD03095047