
Titanium(III) chloride 20% (w/v) in hydrochloric acid 2M, AcroSeal®
Catalog # 429151000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Dark purple Solution
- Titration:18 to 23 %
- Titration with NaOH:≥95 %
Specification Test Results
About this item
:
:154.24 g/mol
Storage Temperature:Ambient
UN:3264
ADR:8,II
:GHS05
CAS Number:7705-07-9
Formula:Cl₃Ti
MDL Number:MFCD00011266