Order Entry
Switzerland
ContactUsLinkComponent
Potassium methanolate 0.1 N in toluene/methanol, AcroSeal®, pure
Potassium methanolate 0.1 N in toluene/methanol, AcroSeal®, pure
Catalog # 429101000.
CAS Number:  
undefined
Potassium methanolate 0.1 N in toluene/methanol, AcroSeal®, pure
Catalog # 429101000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Strength:
    0.095 to 0.105 N
  • Titration Complexometric:
    23.5 to 26.0 % (Zinc)

Specification Test Results

About this item

Synonyms: Potassium methoxide, Potassium methylate
 :
 :70.13 g/mol
Density:0.85 g/cm³
Flash Pt:7 °C
Storage Temperature:Ambient
UN:1992
ADR:3,II
 :GHS06
 :GHS08
 :GHS02
CAS Number:865-33-8
Formula:CH₃KO
MDL Number:MFCD00012416