Order Entry
Switzerland
ContactUsLinkComponent
Bis(2-methoxyethyl)aminosulphur trifluoride 50% (w/w) 2.7 M in toluene, AcroSeal®
Toxic
Health
Corrosive
Flammable
Bis(2-methoxyethyl)aminosulphur trifluoride 50% (w/w) 2.7 M in toluene, AcroSeal®
Catalog # 428981000.
CAS Number:  
undefined
Toxic
Health
Corrosive
Flammable
Bis(2-methoxyethyl)aminosulphur trifluoride 50% (w/w) 2.7 M in toluene, AcroSeal®
Catalog # 428981000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Dark yellow to orange Liquid
  • NMR:
    ≥95 % (On neat substance)
  • Titration with HClO4:
    ≥50 %
  • GC:
    ≥97.0 %

Specification Test Results

About this item

 :
 :221.24 g/mol
Density:1.2 g/cm³
Flash Pt:4 °C
Storage Temperature:Refrigerator
UN:3286
ADR:3,II
 :GHS06
 :GHS08
 :GHS05
 :GHS02
CAS Number:202289-38-1
Formula:C₆H₁₄F₃NO₂S
MDL Number:MFCD01321415