


Ethylene glycol monomethyl ether (2-methoxyethanol) for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-86-4
:
Formula:C₃H₈O₂
MDL Number:MFCD00002867
:76.1 g/mol
Boiling Pt:124 °C (760 mmHg)
Melting Pt:–85 °C
Density:0.96 g/cm³
Flash Pt:38 °C
UN:1188
ADR:3,III
:GHS07
:GHS08
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 %
- GC:≥99.0 %
- UV:1 cm cell vs HPLC-grade water
- Residue after evaporation:≤0.001 %
Specification Test Results
Frequently Bought Together