Dipropyleneglycol monomethyl ether (mixture of isomers) 99%, pure
:34590-94-8
:
:C₇H₁₆O₃
:MFCD00059604
:180 °C (760 mmHg)
:–80 °C
:0.95 g/cm³
:75 °C
:148.2001 g/mol
:Ambient
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥98.5 % sum of isomers
- Elemental Analysis:≥98.5 %
Frequently Bought Together