Cyclohexanone 99+% ACS
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Boiling Pt:155 °C (760 mmHg)
- Density:0.947 g/cm³
- Storage Temperature:Ambient
- UN:1915
- ADR:3,III
3 Options Available Below
Product Details & Documents
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Boiling Pt:155 °C (760 mmHg)
- Density:0.947 g/cm³
- Storage Temperature:Ambient
- UN:1915
- ADR:3,III
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 406092500.
- 406090010.
- 406090025.
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Chem21 Solvent Selection Guide recommended by default
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.406092500.Environmentally PreferableEnvironmental certification - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.406090010.Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationEPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussionSafer Human and Environmental Health - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.406090025.Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended by defaultSafer Human and Environmental Health
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



