


Isobutanol, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-83-1
:
Formula:(CH₃)₂CHCH₂OH
MDL Number:MFCD00004740
:74.12 g/mol
Boiling Pt:108 °C (760 mmHg)
Melting Pt:–108 °C
Density:0.8 g/cm³
Flash Pt:28 °C
UN:1212
ADR:3,III
:GHS07
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.0 %
- Residue after evaporation:≤0.001 %
Specification Test Results
Related Information
Frequently Bought Together