Order Entry
Switzerland
ContactUsLinkComponent
GMBS (N-Succinimidyl 4-maleimidobutyrate) 97%
Irritant
GMBS (N-Succinimidyl 4-maleimidobutyrate) 97%
Catalog # ACRO398681000
CAS Number:  
undefined
Irritant
GMBS (N-Succinimidyl 4-maleimidobutyrate) 97%
Catalog # ACRO398681000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    123 to 130 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :280.24 g/mol
Melting Pt:124…132 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:80307-12-6
Formula:C₁₂H₁₂N₂O₆
MDL Number:MFCD00036817