5-Methoxy-o-toluidine 97%
Catalog # ACRO398240050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to grey to brown Crystalline powder or low melting solid
- Infrared spectrum:Conforms
- Melting point:42 to 46 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
:137.18 g/mol
Boiling Pt:253 °C (1013 hPa)
Melting Pt:43…46 °C
Density:1,39 g/cm³ (25 °C)
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
CAS Number:50868-72-9
Formula:C₈H₁₁NO
MDL Number:MFCD00075057