


Cyclopentane, extra dry 95+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:287-92-3
:
Formula:C₅H₁₀
MDL Number:MFCD00001356
Boiling Pt:50 °C (760 mmHg)
Melting Pt:–94 °C
Density:0.75 g/cm³
Flash Pt:–7 °C
UN:1146
ADR:3,II
:70.13 g/mol
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥95.0 %
- Residue after evaporation:≤0.005 %
Specification Test Results
Related Information
Frequently Bought Together