ContactUsLinkComponent
Dimethyl sulphoxide 99.8+% for HPLC
Dimethyl sulphoxide 99.8+% for HPLC
   Sustainable Options Available
Certificates
undefined
Dimethyl sulphoxide 99.8+% for HPLC
   Sustainable Options Available
Certificates
Specifications
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.1 %
  • GC:
    ≥99.80 %
  • UV:
    1 cm cell vs HPLC-grade water
  • Residue after evaporation:
    ≤0.001 %