Order Entry
Switzerland
ContactUsLinkComponent
4-Amino-m-anisic acid 98%
4-Amino-m-anisic acid 98%
Certificates
undefined
4-Amino-m-anisic acid 98%

Specification Test Results

  • Appearance:
    White to light beige to grey Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    186 to 191 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together