Order Entry
Switzerland
ContactUsLinkComponent
Dibutyltin dilaurate 94%
Corrosive
Irritant
Health
Environment
Dibutyltin dilaurate 94%
Catalog # ACRO382690050
CAS Number:  
undefined
Corrosive
Irritant
Health
Environment
Dibutyltin dilaurate 94%
Catalog # ACRO382690050
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear light yellow Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4690 to 1.4730 (20°C, 589 nm)
  • Tin (Sn):
    17.5 to 19.9 %

Specification Test Results

About this item

CAS Number:77-58-7
 :
Formula:C₃₂H₆₄O₄Sn
MDL Number:MFCD00008963
 :631.57 g/mol
Boiling Pt:205 °C (12 mmHg)
Melting Pt:16…18 °C
Density:1.05 g/cm³
Flash Pt:149 °C
UN:3265
ADR:8,III
 :GHS05
 :GHS07
 :GHS08
 :GHS09
Storage Temperature:Ambient