




Di-n-butylmagnesium 0.5 M in Heptan, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1191-47-5
:
Formula:C₈H₁₈Mg
MDL Number:MFCD00015225
Melting Pt:–71 °C
Density:0,73 g/cm³
Flash Pt:–4 °C
Storage Temperature:Raumtemperatur
UN:3399
ADR:4.3,I
:138.537 g/mol
:GHS07
:GHS08
:GHS05
:GHS09
:GHS02
Specification Test Results
- Appearance:Clear to slightly hazy colorless Solution
- Titration:9.1 to 10.8 % (on Mg basis)
Specification Test Results
Related Information
Frequently Bought Together