SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Chromium
Chromium 99+%, powder, pure
Chromium 99+%, powder, pure
:
Thermo Fisher Scientific
Chromium 99+%, powder, pure
:
Thermo Fisher Scientific
:
7440-47-3
:
:
Cr
:
MFCD00010944
:
2642 °C (760 mmHg)
:
1857 °C
:
7.2 g/cm³
:
Ambient
:
9,III
:
52 g/mol
Appearance:
Grey Metallic powder
Titration with Na2S2O3:
≥99.0 %
ACRO22549
Frequently Bought Together
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
BUFFER PH 10 (500ML) 1 * 3 items
: HAMILTON BONADUZ
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Zinc sulphate heptahydrate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Sodium thiosulphate, Supelco®
: Merck
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection