

Antimony(III) potassium oxitartrate trihydrate ACS
Catalog # ACRO212801000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Titration Iodometric:99.0 to 103.0 %
- Loss on drying:≤2.7 %
- Arsenic (As):≤0.015 %
- Titrable acid or base:≤0.020 meq/g
Specification Test Results
About this item
CAS Number:28300-74-5
:304-61-0
Formula:C₈H₄K₂O₁₂Sb₂·3H₂O
MDL Number:MFCD00167056
:667.88 g/mol
Melting Pt:100 °C
Density:2,6 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1551
ADR:6.1,III
:GHS07
:GHS09