Silver sulphate ACS
: Disilver sulphate
- :10294-26-5
- :Ag₂SO₄
- :MFCD00003407
- :311.8 g/mol
- :1085 °C (760 mmHg)
- :652 °C
- :5,45 g/cm³ (20 °C)
- :Ambient
- :3077
- :9,III
: Disilver sulphate
- :10294-26-5
- :Ag₂SO₄
- :MFCD00003407
- :311.8 g/mol
- :1085 °C (760 mmHg)
- :652 °C
- :5,45 g/cm³ (20 °C)
- :Ambient
- :3077
- :9,III
- Appearance:White Crystalline powder
- Titration Argentometric:≥98.0 %
- Nitrate (NO3):≤0.001 %
- Iron (Fe):≤0.001 %
- Insoluble matter:≤0.02 % (in water and AgCl)
- Subst not pptd by HCl:≤0.03 %
Product Family Options
- Pack typePk



