
Ammonium hexafluorosilicate 99.999% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16919-19-0
:
Formula:H₈F₆N₂Si
MDL Number:MFCD00010887
Density:2.01 g/cm³
UN:2854
ADR:6.1,III
:178.15 g/mol
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder and/or chunks
- Total trace metal impurities:≤10 ppm
Specification Test Results
Frequently Bought Together