Calcium carbonate 99+% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:471-34-1
:
Formula:CaCO₃
MDL Number:MFCD00010906
:100.09 g/mol
Melting Pt:825 °C (dec.)
Density:2,71 g/cm³ (20 °C)
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Titration Complexometric:≥99.0 %
- Chloride (Cl):≤0.005 %
- Sulfate (SO4):≤0.02 %
- Aluminium (Al):≤0.005 %
- Arsenic (As):≤0.0004 %
- Barium (Ba):≤0.005 %
- Iron (Fe):≤0.001 %
- Magnesium (Mg):≤0.05 %
- Potassium (K):≤0.01 %
- Sodium (Na):≤0.2 %
- Strontium (Sr):≤0.1 %
- Zinc (Zn):≤0.001 %
- Heavy metals (as Pb):≤0.002 %
- Insoluble matter:≤0.01 % (in HCl)
Specification Test Results
Frequently Bought Together