



9-BBN monomer 0.5 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:280-64-8
:
Formula:C₈H₁₅B
MDL Number:MFCD00074742
Density:0.884 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:3399
ADR:4.3,II
:122.02 g/mol
:GHS02
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to yellow Liquid (solution under an inert atmosphere)
- Strength:0.45 to 0.55 M (calc. molarity of 9-BBN based on active hydrogen)
Specification Test Results
Related Information
Frequently Bought Together