


m-Xylene, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-38-3
:
Formula:C₈H₁₀
MDL Number:MFCD00008536
:106.17 g/mol
Boiling Pt:139 °C (760 mmHg)
Melting Pt:–48 °C
Density:0.864 g/cm³
Flash Pt:25 °C
UN:1307
ADR:3,III
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.005 %
- GC:≥98.5 %
- Residue after evaporation:≤0.01 %
- Heavy metals (as Pb):≤20 ppm
Specification Test Results
Related Information
Frequently Bought Together