Quartz wool
Marketsource
Supplier: Roth Carl
Jump to:
About this item
This quartz wool is ideal for elementary analysis, as a final stopper in chromatography columns or as a filter aid.
- Chemically pure
- Fibre size 5 to 30 µm
Packaging: Packed in plastic bottles.
2 Options Available Below
Product Details & Documents
This quartz wool is ideal for elementary analysis, as a final stopper in chromatography columns or as a filter aid.
- Chemically pure
- Fibre size 5 to 30 µm
Packaging: Packed in plastic bottles.
Product Family Options
Product Information
- AvailabilityPrice
- ROTH9208.1Quartz woolSpecifications:MarketsourceMarketsource
- ROTH9208.2Quartz woolSpecifications:MarketsourceMarketsource