ContactUsLinkComponent
Laboratory glass cleaner borer 16 PLUS
Laboratory glass cleaner borer 16 PLUS
Catalog # DECO509700.00-K05W
Supplier:  Borer Chemie
undefined
Laboratory glass cleaner borer 16 PLUS
Catalog # DECO509700.00-K05W
Supplier:  Borer Chemie
Specifications
  • Description:
    borer 16 PLUS
  • Seq:
    31
  • Pk:
    5 kg
Specifications
About this item
Liquid, pH-neutral cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials for use in ultrasonic and soaking baths.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • Perfect results even at low concentrations
  • Working solution suitable for multiple use
  • Good material compatibility, especially recommended for alkali and corrosion sensitive laboratory material, also suitable for aluminium
  • Phosphate-free, no release of phosphates into waste water
  • EDTA-, NTA- and chlorine-free, environmentally friendly