Order Entry
Switzerland
ContactUsLinkComponent
VWR®, Taq DNA polymerase, 500 U
VWR®, Taq DNA polymerase, 500 U
Catalog # N224-500U
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Taq DNA polymerase, 500 U
Catalog # N224-500U
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    500
  • Description:
    Taq DNA Polymerase, 500 U

Specifications

About this item

Taq DNA Polymerase is ideal for general PCR and primer extension reactions to amplify DNA templates with high sensitivity. PCR can be initiated from as few as 6 primed templates. Simply add 0,5 µl of Taq DNA Polymerase to a 25 µl PCR reaction.

  • Highly purified, DNA-free, and free of bacterial genomic DNA
  • Amplify DNA templates with high sensitivity
  • Amplify single-copy and low abundance templates

One unit of activity incorporates 10 nmoles of dNTP into acid-insoluble DNA in 30 minutes at 72 °C.

The PCR kit with Taq DNA Polymerase includes separate solutions of all material needed for a PCR reaction, including dNTPs, reaction buffer with and without MgCl2, 25 mM MgCl2, Taq Polymerase, and sterile water.

Delivery: Contains Taq DNA Polymerase, 500 U (5 U/µl) and Taq Buffer, 10X, 1 ml. Sufficient for 200× 25 µl amplifications.